Assay Details
| Gene Symbol: SLC25A38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147518.1 | XP_025003286.1 | 6 | 3667 | 118 | |
| XM_418818.5 | XP_418818.1 | 7 | 3569 | 118 | ||
| GenBank mRNA | CR407387.1 | - | 1 | 914 | 118 | |
More Information
Set Membership: |
|

SLC25A38
Genomic Map