Assay Details
| Gene Symbol: HNF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642413.4 | XP_003642461.1 | 9 | 2821 | 114 | |
| XM_003642414.4 | XP_003642462.1 | 9 | 2743 | 114 | ||
| GenBank mRNA | CR522978.1 | - | 1 | 1116 | 114 | |
More Information
Set Membership: |
|

HNF1B
Genomic Map