Assay Details
| Gene Symbol: LOC418108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416342.6 | XP_416342.1 | 6 - 7 | 804 | 87 | |
| GenBank mRNA | CR523020.1 | - | 6 - 7 | 804 | 87 | |
More Information
Set Membership: |
|

LOC418108
Genomic Map