Assay Details
| Gene Symbol: FAM134A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422050.6 | XP_422050.6 | 9 | 2207 | 95 | |
| GenBank mRNA | CR523026.1 | - | 1 | 923 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM134A| Gene Symbol: FAM134A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422050.6 | XP_422050.6 | 9 | 2207 | 95 | |
| GenBank mRNA | CR523026.1 | - | 1 | 923 | 95 | |
Set Membership: |
|