Assay Details
| Gene Symbol: ACER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004948751.2 | XP_004948808.1 | 5 - 6 | 732 | 52 | |
| XM_418208.6 | XP_418208.2 | 5 - 6 | 723 | 52 | ||
| GenBank mRNA | CR523187.1 | - | 1 - 2 | 50 | 52 | |
More Information
Set Membership: |
|

ACER1
Genomic Map