Assay Details
| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414938.6 | XP_414938.2 | 4 | 1574 | 100 | |
| GenBank mRNA | CR523188.1 | - | 1 | 723 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CARHSP1| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414938.6 | XP_414938.2 | 4 | 1574 | 100 | |
| GenBank mRNA | CR523188.1 | - | 1 | 723 | 100 | |
Set Membership: |
|