Assay Details
| Gene Symbol: LIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294916.2 | XP_015150402.1 | 3 | 106 | ||
| XM_425293.6 | XP_425293.4 | 3 | 106 | |||
| GenBank mRNA | CR523236.1 | - | 1 | 124 | 106 | |
More Information
Set Membership: |
|

LIF
Genomic Map