Assay Details
| Gene Symbol: ZDHHC14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232231.5 | XP_001232232.1 | 5 - 6 | 1637 | 70 | |
| XM_004935655.3 | XP_004935712.1 | 5 - 6 | 1649 | 70 | ||
| XM_004935656.3 | XP_004935713.1 | 5 - 6 | 1649 | 70 | ||
| XM_004935657.3 | XP_004935714.1 | 5 - 6 | 1550 | 70 | ||
| XM_015284228.2 | XP_015139714.1 | 4 - 5 | 1511 | 70 | ||
| XM_025148854.1 | XP_025004622.1 | 5 - 6 | 1538 | 70 | ||
| XM_419692.6 | XP_419692.1 | 5 - 6 | 1637 | 70 | ||
| GenBank mRNA | CR523263.1 | - | 1 - 2 | 65 | 70 | |
More Information
Set Membership: |
|

ZDHHC14
Genomic Map