Assay Details
| Gene Symbol: EHHADH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422690.6 | XP_422690.2 | 6 - 7 | 939 | 78 | |
| GenBank mRNA | CR523580.1 | - | 1 - 2 | 74 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EHHADH| Gene Symbol: EHHADH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422690.6 | XP_422690.2 | 6 - 7 | 939 | 78 | |
| GenBank mRNA | CR523580.1 | - | 1 - 2 | 74 | 78 | |
Set Membership: |
|