Assay Details
| Gene Symbol: NAT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004946352.3 | XP_004946409.1 | 7 | 2135 | 72 | |
| XM_015295473.2 | XP_015150959.1 | 5 | 2163 | 72 | ||
| XM_025141871.1 | XP_024997639.1 | 7 | 1946 | 72 | ||
| XM_420112.6 | XP_420112.2 | 6 | 2262 | 72 | ||
| GenBank mRNA | CR523600.1 | - | 1 | 721 | 72 | |
More Information
Set Membership: |
|

NAT9
Genomic Map