Assay Details
| Gene Symbol: B3GALT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297129.2 | XP_015152615.1 | 1 | 5157 | 93 | |
| GenBank mRNA | CR523844.1 | - | 1 | 625 | 93 | |
More Information
Set Membership: |
|

B3GALT6
Genomic Map