Assay Details
| Gene Symbol: PARP14L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR523878.1 | - | 5 - 6 | 496 | 94 | |
| Gene Symbol: PARP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289949.2 | XP_015145435.1 | 12 - 13 | 4305 | 94 | |
| XM_015289950.2 | XP_015145436.1 | 10 - 11 | 5225 | 94 | ||
| XM_025152691.1 | XP_025008459.1 | 14 - 15 | 5195 | 94 | ||
| XM_025152692.1 | XP_025008460.1 | 14 - 15 | 5192 | 94 | ||
| XM_025152693.1 | XP_025008461.1 | 14 - 15 | 5064 | 94 | ||
More Information
Set Membership: |
|

PARP14L
Genomic Map