Assay Details
| Gene Symbol: PARP14L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR523878.1 | - | 6 - 7 | 701 | 91 | |
| Gene Symbol: PARP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289949.2 | XP_015145435.1 | 13 - 14 | 4510 | 91 | |
| XM_015289950.2 | XP_015145436.1 | 11 - 12 | 5430 | 91 | ||
| XM_025152691.1 | XP_025008459.1 | 15 - 16 | 5400 | 91 | ||
| XM_025152692.1 | XP_025008460.1 | 15 - 16 | 5397 | 91 | ||
| XM_025152693.1 | XP_025008461.1 | 15 - 16 | 5269 | 91 | ||
More Information
Set Membership: |
|

PARP14L
Genomic Map