Assay Details
| Gene Symbol: TMEM35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642569.4 | XP_003642617.3 | 2 - 3 | 313 | 50 | |
| GenBank mRNA | CR524040.1 | - | 2 - 3 | 303 | 50 | |
More Information
Set Membership: |
|

TMEM35B
Genomic Map