Assay Details
| Gene Symbol: FAM188B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943570.3 | XP_004943627.1 | 1 | 1178 | 70 | |
| XM_004943574.3 | XP_004943631.1 | 1 | 1178 | 70 | ||
| XM_004943575.3 | XP_004943632.1 | 1 | 1178 | 70 | ||
| XM_015291841.2 | XP_015147327.1 | 1 | 1178 | 70 | ||
| XM_025153453.1 | XP_025009221.1 | 1 | 1178 | 70 | ||
| XM_025153454.1 | XP_025009222.1 | 1 | 1178 | 70 | ||
| XM_025153455.1 | XP_025009223.1 | 1 | 1178 | 70 | ||
| XM_025153456.1 | XP_025009224.1 | 1 | 1178 | 70 | ||
| XM_025153457.1 | XP_025009225.1 | 1 | 1178 | 70 | ||
| XM_025153458.1 | XP_025009226.1 | 1 | 1178 | 70 | ||
| XM_025153459.1 | XP_025009227.1 | 1 | 1178 | 70 | ||
| GenBank mRNA | CR524061.1 | - | 1 | 618 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004943570.3, XM_004943574.3, XM_004943575.3, XM_015291841.2, XM_025153453.1, XM_025153454.1, XM_025153455.1, XM_025153456.1, XM_025153457.1, XM_025153458.1, XM_025153459.1 transcripts.
Set Membership: |
|

FAM188B2
Genomic Map