Assay Details
| Gene Symbol: MARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278384.2 | XP_015133870.1 | 1 - 2 | 247 | 119 | |
| GenBank mRNA | CR524179.1 | - | 1 - 2 | 221 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015278384.2 transcripts.
Set Membership: |
|

MARS2
Genomic Map