Assay Details
| Gene Symbol: ENOPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149986.1 | XP_025005754.1 | 2 - 3 | 366 | 94 | |
| XM_420559.6 | XP_420559.5 | 2 - 3 | 243 | 94 | ||
| GenBank mRNA | CR524276.1 | - | 2 - 3 | 176 | 94 | |
More Information
Set Membership: |
|

ENOPH1
Genomic Map