Assay Details
| Gene Symbol: PLA2G15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231518.5 | XP_001231519.1 | 6 | 4519 | 108 | |
| GenBank mRNA | CR524355.1 | - | 1 | 716 | 108 | |
More Information
Set Membership: |
|

PLA2G15
Genomic Map