Assay Details
| Gene Symbol: RASL10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233673.5 | XP_001233674.1 | 4 | 1623 | 63 | |
| GenBank mRNA | CR524364.1 | - | 1 | 525 | 63 | |
More Information
Set Membership: |
|

RASL10B
Genomic Map