Assay Details
Gene Symbol: RHBDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277003.2 | XP_015132489.1 | 6 | 2947 | 98 | |
XM_422616.6 | XP_422616.1 | 7 | 3008 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
