Assay Details
| Gene Symbol: LOC770026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233342.3 | XP_001233343.3 | 3 - 4 | 524 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC770026| Gene Symbol: LOC770026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233342.3 | XP_001233343.3 | 3 - 4 | 524 | 84 | |
Set Membership: |
|