Assay Details
| Gene Symbol: PLD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025150878.1 | XP_025006646.1 | 6 - 7 | 1285 | 77 | |
| XM_421399.6 | XP_421399.4 | 6 - 7 | 838 | 77 | ||
More Information
Set Membership: |
|

PLD4
Genomic Map