Assay Details
| Gene Symbol: LMNAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292030.2 | XP_015147516.1 | 12 | 2989 | 99 | |
| XM_015292031.2 | XP_015147517.1 | 11 | 2918 | 99 | ||
More Information
Set Membership: |
|

LMNAL
Genomic Map