Assay Details
| Gene Symbol: GABRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004944775.3 | XP_004944832.1 | 13 | 3975 | 106 | |
| XM_015293616.2 | XP_015149102.1 | 10 | 3121 | 106 | ||
| XM_025155029.1 | XP_025010797.1 | 12 | 4078 | 106 | ||
| XM_025155030.1 | XP_025010798.1 | 13 | 3890 | 106 | ||
| XM_025155031.1 | XP_025010799.1 | 10 | 3166 | 106 | ||
More Information
Set Membership: |
|

GABRP
Genomic Map