Assay Details
| Gene Symbol: PPM1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641050.4 | XP_003641098.1 | 7 - 8 | 1415 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPM1G| Gene Symbol: PPM1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641050.4 | XP_003641098.1 | 7 - 8 | 1415 | 67 | |
Set Membership: |
|