Assay Details
| Gene Symbol: LRRC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293907.2 | XP_015149393.1 | 5 - 6 | 1093 | 82 | |
| XM_416514.6 | XP_416514.2 | 5 - 6 | 939 | 82 | ||
More Information
Set Membership: |
|

LRRC23
Genomic Map