Assay Details
| Gene Symbol: AADAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940922.3 | XP_004940979.1 | 12 - 13 | 1200 | 67 | |
| XM_004940923.3 | XP_004940980.1 | 12 - 13 | 1278 | 67 | ||
| XM_015285387.2 | XP_015140873.1 | 11 - 12 | 1109 | 67 | ||
| XM_426286.6 | XP_426286.3 | 12 - 13 | 1200 | 67 | ||
More Information
Set Membership: |
|

AADAT
Genomic Map