Assay Details
| Gene Symbol: CDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947552.3 | XP_004947609.1 | 11 - 12 | 1265 | 74 | |
| XM_004947553.3 | XP_004947610.1 | 11 - 12 | 1232 | 74 | ||
| XM_417669.6 | XP_417669.1 | 11 - 12 | 1126 | 74 | ||
| GenBank mRNA | KC886601.2 | - | 11 - 12 | 1137 | 74 | |
| KC886602.2 | - | 11 - 12 | 1137 | 74 | ||
| KC886603.2 | - | 11 - 12 | 1137 | 74 | ||
| KC886604.2 | - | 11 - 12 | 1137 | 74 | ||
More Information
Set Membership: |
|

CDS2
Genomic Map