Assay Details
| Gene Symbol: ARFIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015280929.2 | XP_015136415.1 | 7 - 8 | 962 | 72 | |
| XM_015280930.2 | XP_015136416.1 | 6 - 7 | 809 | 72 | ||
| XM_025146978.1 | XP_025002746.1 | 7 - 8 | 1345 | 72 | ||
More Information
Set Membership: |
|

ARFIP2
Genomic Map