Assay Details
| Gene Symbol: NUP62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420179.6 | XP_420179.4 | 10 - 11 | 1723 | 146 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUP62| Gene Symbol: NUP62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420179.6 | XP_420179.4 | 10 - 11 | 1723 | 146 | |
Set Membership: |
|