Assay Details
| Gene Symbol: DLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935407.3 | XP_004935464.1 | 6 | 2407 | 56 | |
| XM_015283772.2 | XP_015139258.1 | 6 | 2226 | 56 | ||
| XM_015283773.1 | XP_015139259.1 | 6 | 2230 | 56 | ||
More Information
Set Membership: |
|

DLK2
Genomic Map