Assay Details
| Gene Symbol: SLC15A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415099.6 | XP_415099.3 | 4 - 5 | 1206 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC15A4| Gene Symbol: SLC15A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415099.6 | XP_415099.3 | 4 - 5 | 1206 | 80 | |
Set Membership: |
|