Assay Details
| Gene Symbol: USP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278916.2 | XP_015134402.1 | 9 | 3564 | 108 | |
| XM_025146840.1 | XP_025002608.1 | 10 | 4586 | 108 | ||
| XM_025146841.1 | XP_025002609.1 | 10 | 4681 | 108 | ||
More Information
Set Membership: |
|

USP12
Genomic Map