Assay Details
| Gene Symbol: MUTYH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936749.3 | XP_004936806.2 | 8 - 9 | 1146 | 58 | |
| XM_015291279.2 | XP_015146765.1 | 8 - 9 | 1147 | 58 | ||
| XM_025153085.1 | XP_025008853.1 | 8 - 9 | 1123 | 58 | ||
More Information
Set Membership: |
|

MUTYH
Genomic Map