Assay Details
| Gene Symbol: TMEM163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289925.2 | XP_015145411.1 | 3 - 4 | 462 | 86 | |
| XM_422132.6 | XP_422132.2 | 4 - 5 | 582 | 86 | ||
More Information
Set Membership: |
|

TMEM163
Genomic Map