Assay Details
| Gene Symbol: RBM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004946908.3 | XP_004946965.1 | 3 - 4 | 435 | 63 | |
| XM_025142285.1 | XP_024998053.1 | 2 - 3 | 86 | 63 | ||
| XM_425690.6 | XP_425690.2 | 3 - 4 | 406 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025142285.1 transcripts.
Set Membership: |
|

RBM23
Genomic Map