Assay Details
Gene Symbol: ADAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288469.2 | XP_015143955.1 | 7 - 8 | 1448 | 112 | |
XM_015288479.2 | XP_015143965.1 | 7 - 8 | 1431 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
