Assay Details
| Gene Symbol: ZBTB24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284564.2 | XP_015140050.1 | 7 | 3203 | 66 | |
| XM_015284565.2 | XP_015140051.1 | 7 | 3000 | 66 | ||
| XM_419793.6 | XP_419793.4 | 7 | 3174 | 66 | ||
More Information
Set Membership: |
|

ZBTB24
Genomic Map