Assay Details
| Gene Symbol: C2H18ORF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419161.6 | XP_419161.4 | 17 - 18 | 3927 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2H18ORF8| Gene Symbol: C2H18ORF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419161.6 | XP_419161.4 | 17 - 18 | 3927 | 90 | |
Set Membership: |
|