Assay Details
| Gene Symbol: SHC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233681.5 | XP_001233682.3 | 11 - 12 | 1828 | 101 | |
| XM_015300005.2 | XP_015155491.1 | 11 - 12 | 1868 | 101 | ||
| XM_015300006.2 | XP_015155492.1 | 11 - 12 | 1868 | 101 | ||
More Information
Set Membership: |
|

SHC2
Genomic Map