Assay Details
| Gene Symbol: MKL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937713.3 | XP_004937770.1 | 14 | 5077 | 63 | |
| XM_004937714.2 | XP_004937771.1 | 14 | 4847 | 63 | ||
| XM_025155225.1 | XP_025010993.1 | 14 | 4838 | 63 | ||
| XM_416243.6 | XP_416243.4 | 16 | 5215 | 63 | ||
More Information
Set Membership: |
|

MKL1
Genomic Map