Assay Details
| Gene Symbol: TM2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001235552.5 | XP_001235553.2 | 2 - 3 | 244 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TM2D1| Gene Symbol: TM2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001235552.5 | XP_001235553.2 | 2 - 3 | 244 | 89 | |
Set Membership: |
|