Assay Details
| Gene Symbol: KYAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945859.2 | XP_004945916.2 | 2 - 3 | 1157 | 58 | |
| XM_015279571.2 | XP_015135057.1 | 3 - 4 | 421 | 58 | ||
| XM_025141439.1 | XP_024997207.1 | 3 - 4 | 279 | 58 | ||
| XM_025141440.1 | XP_024997208.1 | 3 - 4 | 243 | 58 | ||
| XM_415485.6 | XP_415485.2 | 3 - 4 | 399 | 58 | ||
| GenBank mRNA | BX930471.2 | - | 3 - 4 | 267 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025141439.1, XM_025141440.1 transcripts.
Set Membership: |
|

KYAT1
Genomic Map