Assay Details
| Gene Symbol: ATPIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX930483.2 | - | 1 | 318 | 103 | |
| BX932729.1 | - | 1 | 310 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATPIF1| Gene Symbol: ATPIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX930483.2 | - | 1 | 318 | 103 | |
| BX932729.1 | - | 1 | 310 | 103 | ||
Set Membership: |
|