Assay Details
| Gene Symbol: CCNI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276438.2 | XP_015131924.1 | 5 - 6 | 721 | 64 | |
| XM_420590.6 | XP_420590.1 | 4 - 5 | 599 | 64 | ||
| GenBank mRNA | BX930648.2 | - | 4 - 5 | 512 | 64 | |
More Information
Set Membership: |
|

CCNI
Genomic Map