Assay Details
| Gene Symbol: RBPJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936138.3 | XP_004936195.1 | 2 - 3 | 2178 | 61 | |
| XM_015285708.2 | XP_015141194.1 | 2 - 3 | 76 | 61 | ||
| XM_015285709.2 | XP_015141195.1 | 2 - 3 | 2201 | 61 | ||
| XM_015285710.2 | XP_015141196.1 | 2 - 3 | 1007 | 61 | ||
| XM_025149850.1 | XP_025005618.1 | 2 - 3 | 61 | 61 | ||
| XM_025149851.1 | XP_025005619.1 | 2 - 3 | 60 | 61 | ||
| XM_025149852.1 | XP_025005620.1 | 3 - 4 | 312 | 61 | ||
| XM_420752.6 | XP_420752.2 | 2 - 3 | 237 | 61 | ||
| GenBank mRNA | BX930660.1 | - | 1 - 2 | 35 | 61 | |
| BX935186.2 | - | 2 - 3 | 106 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025149851.1, XM_025149852.1 transcripts.
Set Membership: |
|

RBPJ
Genomic Map