Assay Details
| Gene Symbol: ZFAND2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945132.3 | XP_004945189.1 | 1 - 2 | 63 | 56 | |
| XM_422896.5 | XP_422896.1 | 1 - 2 | 109 | 56 | ||
| GenBank mRNA | BX930699.1 | - | 1 - 2 | 147 | 56 | |
| BX933168.1 | - | 1 - 2 | 149 | 56 | ||
| BX933229.1 | - | 1 - 2 | 101 | 56 | ||
| CR407356.1 | - | 1 - 2 | 147 | 56 | ||
More Information
Set Membership: |
|

ZFAND2A
Genomic Map