Assay Details
| Gene Symbol: PHGDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422226.6 | XP_422226.4 | 10 - 11 | 1315 | 101 | |
| GenBank mRNA | BX930701.2 | - | 1 - 2 | 138 | 101 | |
More Information
Set Membership: |
|

PHGDH
Genomic Map