Assay Details
| Gene Symbol: GUCY1B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153727.1 | XP_025009495.1 | 4 - 5 | 747 | 156 | |
| GenBank mRNA | BX930852.2 | - | 1 - 2 | 157 | 135 | |
More Information
Set Membership: |
|

GUCY1B4
Genomic Map