Assay Details
| Gene Symbol: TIMM23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421501.6 | XP_421501.2 | 4 - 5 | 372 | 74 | |
| GenBank mRNA | BX930913.1 | - | 4 - 5 | 356 | 74 | |
| BX933189.1 | - | 4 - 5 | 363 | 74 | ||
| BX933971.2 | - | 4 - 5 | 333 | 74 | ||
| BX935539.2 | - | 4 - 5 | 378 | 74 | ||
| BX950355.2 | - | 4 - 5 | 378 | 74 | ||
More Information
Set Membership: |
|

TIMM23B
Genomic Map